Learn More
Cayman Chemical Dimaprit (hydrochloride), Purity: ≥95%, CAS: 23256-33-9, 50 mg
SDP
Catalog No. 502989083
Encompass_Preferred
A histamine H2 receptor agonist (Ki = 44 µM in guinea pig right atrium); selectively stimulates H2 over H1 and H3 receptors with relative potencies of 71, A. suum extract, which is not affected by pretreatment with mepyramine or cimetidine; stimulates gastric acid secretion in vivo in anaesthetized rats and cats with relative potencies of 19.5 and approximately 400 to 500, respectively, compared to histamine; inhibits 2-(2-pyridyl)ethylamine-induced bronchoconstriction in guinea pigs in a dose-dependent manner
Catalog No. 50-298-9083 Supplier Cayman Chemical Supplier No. 2941850 MG
A histamine H2 receptor agonist (Ki = 44 µM in guinea pig right atrium); selectively stimulates H2 over H1 and H3 receptors with relative potencies of 71, A. suum extract, which is not affected by pretreatment with mepyramine or cimetidine; stimulates gastric acid secretion in vivo in anaesthetized rats and cats with relative potencies of 19.5 and approximately 400 to 500, respectively, compared to histamine; inhibits 2-(2-pyridyl)ethylamine-induced bronchoconstriction in guinea pigs in a dose-dependent manner