Learn More
Medchemexpress LLC Dimboa | 15893-52-4 | 100 MG
SDP
Catalog No. 5000417458
Encompass_Preferred
DIMBOA is a benzoxazinoid found in graminaceous plants such as maize, wheat, and rye, acting as part of their chemical defense system. It exhibits growth inhibitory properties against various bacteria and fungi, including Staphylococcus aureus, Escherichia coli, and Saccharomyces cerevisiae, as well as the mycotoxin-producing fungus Fusarium graminearum.
  • Demonstrates growth inhibitory properties against various bacteria and fungi.
  • Inhibits the mycotoxin-producing fungus Fusarium graminearum.
  • Possesses potent free-radical scavenging activity.
  • Exhibits weaker iron(III) ion reducing activity.
  • Shows overall antioxidant activity.
  • Inhibits the biosynthesis and accumulation of toxic trichothecenes.
  • Reduces plant susceptibility to scab.
  • Exhibits cytotoxicity to plant cells, leading to plasmolysis, cell collapse, and cell rupture.
Catalog No. 50-004-17458 Supplier Medchemexpress LLC Supplier No. HYN7432100MG
DIMBOA is a benzoxazinoid found in graminaceous plants such as maize, wheat, and rye, acting as part of their chemical defense system. It exhibits growth inhibitory properties against various bacteria and fungi, including Staphylococcus aureus, Escherichia coli, and Saccharomyces cerevisiae, as well as the mycotoxin-producing fungus Fusarium graminearum.
  • Demonstrates growth inhibitory properties against various bacteria and fungi.
  • Inhibits the mycotoxin-producing fungus Fusarium graminearum.
  • Possesses potent free-radical scavenging activity.
  • Exhibits weaker iron(III) ion reducing activity.
  • Shows overall antioxidant activity.
  • Inhibits the biosynthesis and accumulation of toxic trichothecenes.
  • Reduces plant susceptibility to scab.
  • Exhibits cytotoxicity to plant cells, leading to plasmolysis, cell collapse, and cell rupture.