Learn More
Medchemexpress LLC Dimedone (5,5-dimethylcyclohexane-1,3-dione) | 126-81-8 | MFCD00001588 | 99.7% | 140.18 g/mol | C8H12O2 | 100 G
SDP
Catalog No. 5000300677
Encompass_Preferred
Dimedone (5,5-dimethylcyclohexane-1,3-dione) is an organic biochemical reagent used in laboratory research. It is commonly employed as a nucleophilic trapping agent for sulfenic acids and as a building block in organic synthesis. Supplied as a high-purity solid, it is intended for research use and analytical characterization.
Catalog No. 50-003-00677 Supplier Medchemexpress LLC Supplier No. HYW015740100G
Dimedone (5,5-dimethylcyclohexane-1,3-dione) is an organic biochemical reagent used in laboratory research. It is commonly employed as a nucleophilic trapping agent for sulfenic acids and as a building block in organic synthesis. Supplied as a high-purity solid, it is intended for research use and analytical characterization.