Learn More
Medchemexpress LLC Dimedone (5,5-dimethylcyclohexane-1,3-dione) | 126-81-8 | MFCD00001588 | 99.7% | 140.18 g/mol | C8H12O2 | 250 G
SDP
Catalog No. 5000302353
Encompass_Preferred
Dimedone (5,5-dimethylcyclohexane-1,3-dione) is an organic biochemical reagent used in life-science research. It functions as a selective probe for sulfenic acid residues and as a building block in organic synthesis. The material is supplied as a high-purity solid suitable for analytical and synthetic applications.
  • High purity (99.7%) for reproducible results.
  • Selective reagent for sulfenic acid detection.
  • Suitable as a synthetic intermediate in organic chemistry.
  • Available in laboratory-scale package sizes for research use.
  • Stable solid form with defined molecular weight (140.18 g/mol).
Catalog No. 50-003-02353 Supplier Medchemexpress LLC Supplier No. 250GHYW015740
Dimedone (5,5-dimethylcyclohexane-1,3-dione) is an organic biochemical reagent used in life-science research. It functions as a selective probe for sulfenic acid residues and as a building block in organic synthesis. The material is supplied as a high-purity solid suitable for analytical and synthetic applications.
  • High purity (99.7%) for reproducible results.
  • Selective reagent for sulfenic acid detection.
  • Suitable as a synthetic intermediate in organic chemistry.
  • Available in laboratory-scale package sizes for research use.
  • Stable solid form with defined molecular weight (140.18 g/mol).