Learn More
Medchemexpress LLC HY-B2215 50mg Medchemexpress, Dimemorfan (phosphate) CAS:36304-84-4 Purity:>98%
SDP
Catalog No. 501933111
Encompass_Preferred
Medchemexpress, HY-B2215 50mg Dimemorfan (phosphate) CAS:36304-84-4 Dimemorfan phosphate is a sigma 1 receptor agonist, used as a potent antitussive. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3111 Supplier Medchemexpress LLC Supplier No. HYB221550MG
Medchemexpress, HY-B2215 50mg Dimemorfan (phosphate) CAS:36304-84-4 Dimemorfan phosphate is a sigma 1 receptor agonist, used as a potent antitussive. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.