Learn More
Apexbio Technology LLC DIMENHYDRINATE-200MG
SDP
Catalog No. 5000568663
Encompass_Preferred
Dimenhydrinate (CAS No 523-87-5) is a compound frequently studied in the field of pharmacology for its properties as an antihistamine and antiemetic agent Originating as a combination of diphenhydramine and 8-chlorotheophylline its mechanism of action involves the antagonism of H1 histamine receptors and modulating neural pathways in the vestibular system providing relief from motion sickness and nausea In vitro activity of dimenhydrinate is observed within the nanomolar to low micromolar range It is commonly utilized in various cell models and animal studies to explore its pharmacokinetics and pharmacodynamics as well as in drug screening applications to evaluate its efficacy and safety profiles Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68663 Supplier Apexbio Technology LLC Supplier No. C6570200
Dimenhydrinate (CAS No 523-87-5) is a compound frequently studied in the field of pharmacology for its properties as an antihistamine and antiemetic agent Originating as a combination of diphenhydramine and 8-chlorotheophylline its mechanism of action involves the antagonism of H1 histamine receptors and modulating neural pathways in the vestibular system providing relief from motion sickness and nausea In vitro activity of dimenhydrinate is observed within the nanomolar to low micromolar range It is commonly utilized in various cell models and animal studies to explore its pharmacokinetics and pharmacodynamics as well as in drug screening applications to evaluate its efficacy and safety profiles Experimental concentrations typically depend on the specific research design