Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimenhydrinate, MP Biomedicals™

Catalog No. ICN19018910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19018910 10 g
ICN19018990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19018910 Supplier MP Biomedicals, Inc Supplier No. 0219018910
Add to Cart
Add to Cart

Chemical Identifiers

CAS 523-87-5
Molecular Formula C24H28ClN5O3
Molecular Weight (g/mol) 469.97
InChI Key NFLLKCVHYJRNRH-UHFFFAOYSA-N
Synonym dimenhydrinate, dramamine, vomex a, chloranautine, diphenhydrinate, anautine, travelin, amosyt, aviomarin, gravol
PubChem CID 10660
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Cl.CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color White
Quantity 10 g
Formula Weight 470
Physical Form Powder
Chemical Name or Material Dimenhydrinate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.