Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimenhydrinate, USP, 53-55.5%, Spectrum™ Chemical
SDP

Catalog No. 18602231 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-231 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-231 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DI12725KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

DI127, 523-87-5, C17H21NO·C7H7ClN4O2

Spectrum™ Chemical Dimenhydrinate, USP is used to prevent and treat nausea, vomiting, and dizziness caused by motion sickness. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 523-87-5
CAS Max % 100%
Ignition Residue 0.3%
Loss on Drying 0.5%
Packaging Fiber Drum
Molecular Formula C24H28ClN5O3
Quantity 25 kg
InChI Key NFLLKCVHYJRNRH-UHFFFAOYSA-N
SMILES CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O.CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione; [2-(diphenylmethoxy)ethyl]dimethylamine
Molecular Weight (g/mol) 469.97
Percent Purity 53 to 55.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Dimenhydrinate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.