Learn More
Siemens Healthineers™ Dimension™ EXL™ 200 RMS Integrated Chemistry System with LM Export
Catalog No. 23313013 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sample Type:
Blood
Catalog No. Sample Type
23-313-013 Blood
1 options
This item is not returnable. View return policy
Catalog No. 23-313-013 Supplier Siemens Healthineers™ Supplier No. 10472176
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
Simplify your operations with this integrated clinical chemistry and immunoassay analyzer.
The Dimension™ EXL™ 200 Integrated Chemistry System, now including Dimension Remote Services and enhanced QCC PowerPak™, gives you the power of both chemistry and immunoassay testing in a compact system ideal for smaller-sized laboratories.
  • Offers versatility to deliver better outcomes in any location with a comprehensive menu, including critical cardiac assays and our automated-pretreatment immunosuppressive drug assays
  • Mix and match chemistry and immunoassay tests on the fly in one common reagent
  • Transition easily and correlate results between core and satellite labs using the partnership of the Atellica™ Solution and the Dimension EXL 200 Integrated Chemistry System
  • High productivity and STAT sampling from any position
  • Avoids splitting samples by allowing you to run all assays from one sample tube
  • 4 min. to first result for a basic metabolic panel; electrolyte results in less than 1 min.
  • Same time with no manual sample pretreatment, no manual regent preparation, and log calibration stability
TRUSTED_SUSTAINABILITY
Specifications
Type Analyzer
Sample Type Blood
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov