Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimer acid, hydrogenated

Catalog No. 4323691L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-236-91L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-236-91L Supplier Sigma Aldrich Supplier No. 4323691L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Improves flexibility, toughness, impact resistance, dyeability and hydrolytic stability of polymers.Dimer acid, hydrogenated can be used as a corrosion inhibitor for the protection of lead metal substrates. It can also be used in polyamide based hot melting adhesives as a thermoplastic polymer for electronic boards, textiles and packaging systems.

Specifications

CAS 68783-41-5
Density 0.95 g/mL (at 25°C)
Refractive Index n20/D 1.477 (literature)
MDL Number MFCD00211870
Quantity 1 L
Synonym Fatty acids, C18 -unsatd., dimers, hydrogenated; Hydrogenated C36 dimer fatty acid
Molecular Weight (g/mol) 570
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.