Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimercaprol | 59-52-9 | 98.0% | 250 MG
SDP

Catalog No. 5000413742
Encompass_Preferred

Dimercaprol (2,3-Dimercapto-1-propanol) is an anti-heavy metal-poisoning agent that also exhibits anti-HIV activity. It is used for the study of poisoning by arsenic, mercury, gold, lead, antimony, and other toxic metals.

  • Anti-heavy metal-poisoning agent
  • Exhibits anti-HIV activity
  • Liquid appearance with a density of 1.239 g/cm³
  • Colorless to off-white
  • Molecular weight of 124.23
  • Formula C3H8OS2
  • Storage at 4°C, stored under nitrogen
  • For research use only
  • Can be used to study arsenic, mercury, gold, lead, antimony, and other toxic metal poisoning
  • Protects PC-12 cells from Acrolein-mediated cell death in a dose-dependent manner in vitro
  • Decreases mercury content in the liver and kidney in Wistar rats exposed to HgCl2 in vivo

Catalog No. 50-004-13742 Supplier Medchemexpress LLC Supplier No. 250MGHYB1285
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimercaprol (2,3-Dimercapto-1-propanol) is an anti-heavy metal-poisoning agent that also exhibits anti-HIV activity. It is used for the study of poisoning by arsenic, mercury, gold, lead, antimony, and other toxic metals.

  • Anti-heavy metal-poisoning agent
  • Exhibits anti-HIV activity
  • Liquid appearance with a density of 1.239 g/cm³
  • Colorless to off-white
  • Molecular weight of 124.23
  • Formula C3H8OS2
  • Storage at 4°C, stored under nitrogen
  • For research use only
  • Can be used to study arsenic, mercury, gold, lead, antimony, and other toxic metal poisoning
  • Protects PC-12 cells from Acrolein-mediated cell death in a dose-dependent manner in vitro
  • Decreases mercury content in the liver and kidney in Wistar rats exposed to HgCl2 in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.