Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimercaptomaleonitrile disodium salt hydrate

Catalog No. 385105GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-510-5GF 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-510-5GF Supplier Sigma Aldrich Supplier No. 385105GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimercaptomaleonitrile disodium salt hydrate

Specifications

CAS 5466-54-6
Molecular Formula C4N2Na2S2 -+ xH2O
Linear Formula C4N2Na2S2 -+ xH2O
MDL Number MFCD00217055
Quantity 100 mg
Molecular Weight (g/mol) 186.17 (anhydrous basis)
Percent Purity ≥95.0% (calc. based on dry substance, NT); ∼10%crystal water basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.