Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimesitylammonium Pentafluorobenzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. D32931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32931G 1 g
D32935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32931G Supplier TCI America Supplier No. D32931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 850629-65-1
Molecular Formula C24H24F5NO3S
Molecular Weight (g/mol) 501.512
InChI Key MKKHTKBUMNIBLN-UHFFFAOYSA-N
PubChem CID 44629836
IUPAC Name bis(2,4,6-trimethylphenyl)azanium;2,3,4,5,6-pentafluorobenzenesulfonate
SMILES CC1=CC(=C(C(=C1)C)[NH2+]C2=C(C=C(C=C2C)C)C)C.C1(=C(C(=C(C(=C1F)F)S(=O)(=O)[O-])F)F)F

Specifications

Quantity 1 g
Formula Weight 501.51
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimesitylammonium Pentafluorobenzenesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.