Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimesitylcyclopropenone, 99%, Thermo Scientific™

Catalog No. AC407850050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407850050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407850050 Supplier Thermo Scientific Chemicals Supplier No. 407850050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61440-88-8
Molecular Formula C21H22O
Molecular Weight (g/mol) 290.406
InChI Key LXCSCXXFUGZMQN-UHFFFAOYSA-N
Synonym 2,3-dimesitylcycloprop-2-enone, 2,3-bis 2,4,6-trimethylphenyl cycloprop-2-en-1-one, 2-cyclopropen-1-one,2,3-bis 2,4,6-trimethylphenyl, 2,3-dimesityl-2-cyclopropene-1-one
PubChem CID 2733921
IUPAC Name 2,3-bis(2,4,6-trimethylphenyl)cycloprop-2-en-1-one
SMILES CC1=CC(=C(C(=C1)C)C2=C(C2=O)C3=C(C=C(C=C3C)C)C)C

Specifications

Assay Percent Range 99%
Quantity 5 g
Formula Weight 290.4
Chemical Name or Material Dimesitylcyclopropenone, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.