Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimesitylmethane, 98+%, Thermo Scientific™

Catalog No. AAL0109014
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0109014 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0109014 Supplier Thermo Scientific Chemicals Supplier No. L0109014

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 733-07-3
Molecular Formula C19H24
Quantity 25g
MDL Number MFCD00014913
Beilstein 2053472
Synonym dimesitylmethane, 1,1'-methylenebis 2,4,6-trimethylbenzene, 1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl benzene, benzene, 1,1'-methylenebis 2,4,6-trimethyl, benzene,1'-methylenebis 2,4,6-trimethyl, 2-mesitylmethyl-1,3,5-trimethylbenzene, 2-mesitylmethyl-1,3,5-trimethylbenzene #, benzene,1,3,5-trimethyl-2-2,4,6-trimethylphenyl methyl
InChI Key ZQYQPPLKPFBKLD-UHFFFAOYSA-N
SMILES CC1=CC(=C(C(=C1)C)CC2=C(C=C(C=C2C)C)C)C
IUPAC Name 1,3,5-trimethyl-2-[(2,4,6-trimethylphenyl)methyl]benzene
Molecular Weight (g/mol) 252.401
PubChem CID 69771
Formula Weight 252.4
Percent Purity ≥98%
Chemical Name or Material Dimesitylmethane
Show More Show Less
EINECSNumber 211-991-6
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.