Learn More
FUJIFILM Biosciences Dimethenamid Standard
Catalog No. 5000450438
Encompass_Preferred
High-purity Dimethenamid standard 99 0% for pesticide residue analysis compatible with capillary gas chromatography GC
Catalog No. 50-004-50438 Supplier FUJIFILM Biosciences Supplier No. 04326751
High-purity Dimethenamid standard 99 0% for pesticide residue analysis compatible with capillary gas chromatography GC