Learn More
Selleck Chemical LLC Dimethindene maleate-E1796-5MG
SDP
Catalog No. 503139087
Encompass_Preferred
Dimethindene maleate is a selective antagonist of histamine H1 and exhibits antihistamine effects It suppresses the cromakalim-induced/glibenclamide-sensitive K currents in a concentration-dependent and reversible manner
Catalog No. 50-313-9087 Supplier Selleck Chemical LLC Supplier No. E17965MG
Dimethindene maleate is a selective antagonist of histamine H1 and exhibits antihistamine effects It suppresses the cromakalim-induced/glibenclamide-sensitive K currents in a concentration-dependent and reversible manner