Learn More
Cayman Chemical Dimethindene (maleate), Purity: ≥98%, CAS: 3614-69-5, 100 mg
SDP
Catalog No. 502942933
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 1.5 nM); selective for histamine H1 receptors over M1 mAChRs and 5-HT receptor subtype 5-HT2A (Kis = 64 and 2,400 nM, respectively); alone or in combination with icodextrin decreases adhesions between the abdominal wall and intraperitoneal viscera and local vascularization following surgical-induced abdominal trauma in rabbits at 0.1 mg/kg
Catalog No. 50-294-2933 Supplier Cayman Chemical Supplier No. 40649100 MG
A histamine H1 receptor antagonist (Ki = 1.5 nM); selective for histamine H1 receptors over M1 mAChRs and 5-HT receptor subtype 5-HT2A (Kis = 64 and 2,400 nM, respectively); alone or in combination with icodextrin decreases adhesions between the abdominal wall and intraperitoneal viscera and local vascularization following surgical-induced abdominal trauma in rabbits at 0.1 mg/kg