Learn More
Cayman Chemical Dimethoate-d6, Purity: ≥99% deuterated forms (d1-d6), CAS: 1219794-81-6, 10 mg
SDP
Catalog No. 502916565
Encompass_Preferred
An internal standard for the quantification of dimethoate by GC- or LC-MS
Catalog No. 50-291-6565 Supplier Cayman Chemical Supplier No. 3926010 MG
An internal standard for the quantification of dimethoate by GC- or LC-MS