Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethocaine hydrochloride | 553-63-9 | 99.4% | 500 MG
SDP

Catalog No. 5000233708
Encompass_Preferred

Dimethocaine hydrochloride is the hydrochloride salt of dimethocaine (Larocaine). It is supplied for research use and has been reported as a new psychoactive substance (NPS); handle per institutional safety protocols.

  • CAS number: 553-63-9.
  • Purity: 99.36% (as listed).
  • Molecular formula: C16H27ClN2O2.
  • Molecular weight: 314.85 g/mol.
  • Solubility: soluble in DMSO and water.
  • Available package sizes: 25 mg, 50 mg, 100 mg, 500 mg, 1 g, 5 g.

Catalog No. 50-002-33708 Supplier Medchemexpress LLC Supplier No. HY121870A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethocaine hydrochloride is the hydrochloride salt of dimethocaine (Larocaine). It is supplied for research use and has been reported as a new psychoactive substance (NPS); handle per institutional safety protocols.

  • CAS number: 553-63-9.
  • Purity: 99.36% (as listed).
  • Molecular formula: C16H27ClN2O2.
  • Molecular weight: 314.85 g/mol.
  • Solubility: soluble in DMSO and water.
  • Available package sizes: 25 mg, 50 mg, 100 mg, 500 mg, 1 g, 5 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.