Learn More
Cayman Chemical Dimethocaine (hydrochloride), Purity: ≥98%, CAS: 553-63-9, 10 mg
SDP
Catalog No. 502951242
Encompass_Preferred
A local anesthetic that, because of its similarity in action to cocaine, has potential for abuse; completely inhibits dopamine uptake in rat striatal synaptosomes with an IC50 value (1.2 µM) comparable to that of cocaine (IC50 = 0.7 µM); substitutes for cocaine in drug discrimination tests in rats and rhesus monkeys; intended for forensic and research purposes
Catalog No. 50-295-1242 Supplier Cayman Chemical Supplier No. 1115910 MG
A local anesthetic that, because of its similarity in action to cocaine, has potential for abuse; completely inhibits dopamine uptake in rat striatal synaptosomes with an IC50 value (1.2 µM) comparable to that of cocaine (IC50 = 0.7 µM); substitutes for cocaine in drug discrimination tests in rats and rhesus monkeys; intended for forensic and research purposes