Learn More
Cayman Chemical Dimethomorph, Purity: ≥95% (mixture of cis and trans isomers), CAS: 110488-70-5, 100 mg
SDP
Catalog No. 502935692
Encompass_Preferred
A morpholine fungicide; inhibits fungal cell wall formation; inhibits mycelial growth of the oomycete fungi P. citrophthora, P. parasitica, P. capsici, and P. infestans in vitro (EC50s = 0.14, 0.38, C. vulgaris or S. obliquus in vitro (EC50s = 47.46 and 44.87 µg/ml, respectively); inhibits AR activity in a reporter assay in MDA-kb2 human breast cancer cells, but not in a yeast antiandrogen screen (IC20s = 0.263 and 38.5 µM, respectively); not toxic to rats (LD50 = 3,900 mg/kg) or goldfish (C. auratus; LC50 = >32 µg/ml)
Catalog No. 50-293-5692 Supplier Cayman Chemical Supplier No. 24247100 MG
A morpholine fungicide; inhibits fungal cell wall formation; inhibits mycelial growth of the oomycete fungi P. citrophthora, P. parasitica, P. capsici, and P. infestans in vitro (EC50s = 0.14, 0.38, C. vulgaris or S. obliquus in vitro (EC50s = 47.46 and 44.87 µg/ml, respectively); inhibits AR activity in a reporter assay in MDA-kb2 human breast cancer cells, but not in a yeast antiandrogen screen (IC20s = 0.263 and 38.5 µM, respectively); not toxic to rats (LD50 = 3,900 mg/kg) or goldfish (C. auratus; LC50 = >32 µg/ml)