Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethoxydi-p-tolylsilane 98.0+%, TCI America™
SDP

Catalog No. D37335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37335G 5 g
D37331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37335G Supplier TCI America Supplier No. D37335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92779-72-1
Molecular Formula C16H20O2Si
Molecular Weight (g/mol) 272.419
MDL Number MFCD01861714
InChI Key SZIZIGBTHTUEBU-UHFFFAOYSA-N
PubChem CID 11821755
IUPAC Name dimethoxy-bis(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](C2=CC=C(C=C2)C)(OC)OC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 272.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxydi-p-tolylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.