Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethoxydimethylsilane

Catalog No. 104906500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-490-6500ML 500 mL
10-490-61L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 10-490-6500ML Supplier Sigma Aldrich Supplier No. 104906500ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethoxydimethylsilane (DMDMS) can be used:To synthesize various macroporous silica aerogels.In the preparation of hollow silica nanoparticles loaded with phthalocyanine for near-infrared photodynamic and photothermal combination therapy.To synthesize dielectric silicone elastomer with high dielectric permittivity.

Specifications

CAS 1112-39-6
Density 0.88 g/mL at 25°C
Boiling Point 81.4°C
Molecular Formula C4H12O2Si
Linear Formula Si(OCH3)2(CH3)2
Refractive Index n20/D 1.369
MDL Number MFCD00025691
Quantity 500 mL
Synonym DMDMS; Dimethyldimethoxysilane
Molecular Weight (g/mol) 120.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.