Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethoxydiphenylsilane

Catalog No. 42940100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-940-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 42-940-100ML Supplier Sigma Aldrich Supplier No. 42940100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethoxydiphenylsilane (DMDPS, DDS) is an acetylating reagent.{57}DMDPS may be useful as a precursor to prepare silica.{58} It-ßmay be used in lithium ion batteries as an electrolyte additive. It prevents the batteries from overcharging.{59}

Specifications

CAS 879-05-0
Density 0.861 g/mL at 25°C
Boiling Point 34.6°C
Molecular Formula C6BrF5Mg
Refractive Index n20/D 1.5450
Linear Formula C6F5MgBr
MDL Number MFCD00015720
Quantity 100 mL
Synonym Diphenyldimethoxysilane
Molecular Weight (g/mol) 271.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.