Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl (1-Diazo-2-Oxopropyl)Phosphonate Solution

Catalog No. 74272410ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-272-410ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 74-272-410ML Supplier Sigma Aldrich Supplier No. 74272410ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyl (1-diazo-2-oxopropyl)phosphonate can be used as a reagent for the synthesis of: Ethynyl compounds (alkynes) from aldehydes. Isoquinoline and pyridine N-oxides by cyclization with oximes in the presence of Rh catalyst. Dimethyl(diazomethyl)phosphonate through methanolysis, which is further employed as a reagent in the synthesis of enol ethers or alkynes. Five-member heterocyclic scaffolds of pyrazoles, triazoles, and oxazoles via cycloaddition reaction and multicomponent reaction.

Specifications

CAS 1462397-86-9
Melting Point 275°C to 295°C
Molecular Formula C38H36Cl2N2P2Ru
Linear Formula C38H36Cl2N2P2Ru
Refractive Index n20/D 1.352-1.354
MDL Number MFCD26960800
Quantity 500 mg
Synonym Ru-PNN catalyst
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.