Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 2,2'-Bipyridine-4,4'-dicarboxylate 98.0+%, TCI America™
SDP

Catalog No. D46351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46351G 1 g
D46355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46351G Supplier TCI America Supplier No. D46351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71071-46-0
Molecular Formula C14H12N2O4
Molecular Weight (g/mol) 272.26
MDL Number MFCD02669916
InChI Key HBWBVIDKBKOVEX-UHFFFAOYSA-N
PubChem CID 326419
IUPAC Name methyl 2-(4-methoxycarbonylpyridin-2-yl)pyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)C2=NC=CC(=C2)C(=O)OC

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 1 g
Synonym 2,2′C-Bipyridine-4,4′C-dicarboxylic Acid Dimethyl Ester, 4,4′C-Bis(methoxycarbonyl)-2,2′C-bipyridine, 4,4′C-Dimethoxycarbonyl-2,2′C-bipyridine
Formula Weight 272.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,2′-Bipyridine-4,4′-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.