Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Dimethyl 2,3-O-Isopropylidene-D-Tartrate

Catalog No. 3843135G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-431-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-431-35G Supplier Sigma Aldrich Supplier No. 3843135G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate is a chiral reagent used in organic synthesis. It is prepared by the reaction-rectification process from D -tartrate, and 2,2-dimethoxypropane.{138]Valuable building block for TADDOL chiral auxiliaries, dipyridine ligands, and threitols.

Specifications

CAS 337-20-2
Melting Point -126°C
Molecular Formula C4F8O
Refractive Index n20/D 1.439
Linear Formula CF3CF2COCF3
MDL Number MFCD00216737
Quantity 5 g
Synonym Perfluoro-2-butanone
Molecular Weight (g/mol) 216.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.