Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl (-)-2,3-O-Isopropylidene-L-tartrate 95.0+%, TCI America™
SDP

Catalog No. I04475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04475G 5 g
I044725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04475G Supplier TCI America Supplier No. I04475G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37031-29-1
Molecular Formula C9H14O6
Molecular Weight (g/mol) 218.21
MDL Number MFCD00066218
InChI Key ROZOUYVVWUTPNG-PHDIDXHHSA-N
Synonym 4r,5r-dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, --dimethyl 2,3-o-isopropylidene-l-tartrate, dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, --dimethyl-2,3-o-isopropylidene-l-tartrate, 4,5-dimethyl 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, pubchem6766, dimethyl--2,3-o-isopropylidene-l-tartrate, ksc917k0j, rozouyvvwutpng-phdidxhhsa
PubChem CID 688155
IUPAC Name 4,5-dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
SMILES COC(=O)[C@@H]1OC(C)(C)O[C@H]1C(=O)OC

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 g
Formula Weight 218.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (-)-2,3-O-Isopropylidene-L-tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.