Learn More
Dimethyl 2,6-dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylate, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 50428-14-3 |
| Flash Point | Not Applicable |
| Molecular Formula | C17H16N2O5 |
| Linear Formula | C17H16N2O5 |
| Quantity | 25 mg |
| MDL Number | MFCD22572715 |
| Molecular Weight (g/mol) | 328.32 |
| Formula Weight | 328.32 |
| Grade | BP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.