Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 2,7-Naphthalenedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. N06845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06845G 5 g
N06841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06845G Supplier TCI America Supplier No. N06845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2549-47-5
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD03427560
InChI Key WYIBAMPRACRCOM-UHFFFAOYSA-N
Synonym 2,7-Naphthalenedicarboxylic Acid Dimethyl Ester
PubChem CID 9881315
IUPAC Name dimethyl naphthalene-2,7-dicarboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=CC(=C2)C(=O)OC

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 244.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 2,7-Naphthalenedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.