Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl 2-bromo-1H-imidazole-4,5-dicarboxylate

Catalog No. SYX003831G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003831G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003831G Supplier Sigma Aldrich Supplier No. SYX003831G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • One-step preparation of 3-chloroanisole from the corresponding 3-substituted nitrobenzene has been reported.3-chloroanisole was employed as starting reagent in the regioselective synthesis of 4- and 7-alkoxyindoles
  • It was also employed as electrolyte additive for the overcharging protection of li-ion cell

Specifications

CAS 705280-65-5
Molecular Formula C7H7BrN2O4
Linear Formula C7H7BrN2O4
MDL Number MFCD09702026
Quantity 25 g
Molecular Weight (g/mol) 263.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.