Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl (2-Hydroxyethyl)phosphonate, TCI America™
SDP

Catalog No. H07795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07795G 5 g
H077925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07795G Supplier TCI America Supplier No. H07795G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54731-72-5
Molecular Formula C4H11O4P
Molecular Weight (g/mol) 154.10
MDL Number MFCD00043376
InChI Key TZPPDWDHNIMTDQ-UHFFFAOYSA-N
Synonym dimethyl 2-hydroxyethylphosphonate, dimethyl 2-hydroxyethyl phosphonate, 2-hydroxyethylphosphonic acid dimethyl ester, 2-hydroxyethyl phosphonic acid dimethyl ester, acmc-1ao1t, 2-diethylphosphono ethan-1-ol, tzppdwdhnimtdq-uhfffaoysa, hydroxyethyldimethylphosphonic acid, 2-dimethoxyphosphoryl ethan-1-ol
PubChem CID 2733833
IUPAC Name dimethyl (2-hydroxyethyl)phosphonate
SMILES COP(=O)(CCO)OC

Specifications

Color Colorless
Boiling Point 97°C
Quantity 5 g
Formula Weight 154.10
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (2-Hydroxyethyl)phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.