Learn More
Medchemexpress LLC Dimethyl 2-methylidenebutanedioate | 617-52-7 | MFCD00008446 | 99.4% | 158.15 g/mol | C7H10O4 | 100 G
SDP
Catalog No. 5000208797
Encompass_Preferred
Dimethyl 2-methylidenebutanedioate (dimethyl itaconate) is a small-molecule research reagent used to modulate reactive oxygen species and innate immune responses in vitro. It is commonly used in studies of immunomodulation and microglial activation and is supplied with high chemical purity for laboratory research.
  • High chemical purity suitable for in vitro studies.
  • Used as a reactive oxygen species (ROS) modulator and inducer.
  • Applied in immunomodulation and microglial activation research.
  • Molecular formula C7H10O4; molecular weight 158.15 g/mol.
  • Available as a 100 g pack for laboratory use.
Catalog No. 50-002-08797 Supplier Medchemexpress LLC Supplier No. HYW041892100G
Dimethyl 2-methylidenebutanedioate (dimethyl itaconate) is a small-molecule research reagent used to modulate reactive oxygen species and innate immune responses in vitro. It is commonly used in studies of immunomodulation and microglial activation and is supplied with high chemical purity for laboratory research.
  • High chemical purity suitable for in vitro studies.
  • Used as a reactive oxygen species (ROS) modulator and inducer.
  • Applied in immunomodulation and microglial activation research.
  • Molecular formula C7H10O4; molecular weight 158.15 g/mol.
  • Available as a 100 g pack for laboratory use.