Learn More
Medchemexpress LLC Dimethyl 2-oxoglutarate | 13192-04-6 | 97.5% | 174.15 | 5 G
SDP
Catalog No. 5000405120
Encompass_Preferred
Dimethyl 2-oxoglutarate is a cell-permeable derivative of 2-oxoglutarate and a tricarboxylic acid cycle metabolite. It possesses antioxidant properties. It is intended for research use only and is not sold to patients.
  • Cell-permeable derivative
  • Antioxidant properties
  • Inhibits autophagy
  • Prevents mitochondrial damage
  • Reduces ROS production
  • Alleviates liver fibrosis
  • Can be used in the research of diseases such as Alzheimer's disease, diabetes, and cardiomyopathy
Catalog No. 50-004-05120 Supplier Medchemexpress LLC Supplier No. HY441345G
Dimethyl 2-oxoglutarate is a cell-permeable derivative of 2-oxoglutarate and a tricarboxylic acid cycle metabolite. It possesses antioxidant properties. It is intended for research use only and is not sold to patients.
  • Cell-permeable derivative
  • Antioxidant properties
  • Inhibits autophagy
  • Prevents mitochondrial damage
  • Reduces ROS production
  • Alleviates liver fibrosis
  • Can be used in the research of diseases such as Alzheimer's disease, diabetes, and cardiomyopathy