Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl 2-Oxopropylphosphonate

Catalog No. 18069625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-069-625G 25 g
18-069-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-069-625G Supplier Sigma Aldrich Supplier No. 18069625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

Specifications

CAS 4202-14-6
Density 1.202 g/mL at 25°C
Boiling Point 76°C to 79°C (3 mmHg, lit.)
Molecular Formula C5H11O4P
Linear Formula CH3COCH2P(O)(OCH3)2
Refractive Index n20/D 1.439
MDL Number MFCD00008769
Quantity 25 g
Synonym Dimethyl acetonylphosphonate
Molecular Weight (g/mol) 166.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.