Learn More
Crescent Chemical Co Inc DIMETHYL-2-THIAZOLYL)-2 5-DIPH
SDP
Catalog No. 50247525
Encompass_Preferred
(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium-bromide research grade Detection of dehydrogenases in combination with phenazine methosulfate (1). Terminal electron acceptor in the cycling assay for pyridine nucleotides (2). For colorimetric assay to measure cell activation (3, 4). Size - 25G
Catalog No. 50-247-525 Supplier Crescent Chemical Co Inc Supplier No. SE20395.04
(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium-bromide research grade Detection of dehydrogenases in combination with phenazine methosulfate (1). Terminal electron acceptor in the cycling assay for pyridine nucleotides (2). For colorimetric assay to measure cell activation (3, 4). Size - 25G