Learn More
Crescent Chemical Co Inc DIMETHYL-2-THIAZOLYL)-2 5-DIPH
SDP
Catalog No. 50247522
Encompass_Preferred
(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium-bromide research grade Detection of dehydrogenases in combination with phenazine methosulfate (1). Terminal electron acceptor in the cycling assay for pyridine nucleotides (2). For colorimetric assay to measure cell activation (3, 4). Size - 250MG
Catalog No. 50-247-522 Supplier Crescent Chemical Co Inc Supplier No. SE20395.01
(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium-bromide research grade Detection of dehydrogenases in combination with phenazine methosulfate (1). Terminal electron acceptor in the cycling assay for pyridine nucleotides (2). For colorimetric assay to measure cell activation (3, 4). Size - 250MG