Learn More
Crescent Chemical Co Inc DIMETHYL-2-THIAZOLYL)-2 5-DIPH
SDP
Supplier:  Crescent Chemical Co Inc SE20395.03
Encompass_Preferred
(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium-bromide research grade Detection of dehydrogenases in combination with phenazine methosulfate (1). Terminal electron acceptor in the cycling assay for pyridine nucleotides (2). For colorimetric assay to measure cell activation (3, 4). Size - 5G S
Catalog No. 50-247-524