Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl 3,3'-disulfanediyl(2R,2'R)-bis(2-aminopropanoate) dihydrochloride | 32854-09-4 | ≥98.0% | 341.28 | 25 G
SDP

Catalog No. 5000366436
Encompass_Preferred

Dimethyl 3,3'-disulfanediyl(2R,2'R)-bis(2-aminopropanoate) dihydrochloride is a cysteine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 175 mg/mL (512.78 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Catalog No. 50-003-66436 Supplier Medchemexpress LLC Supplier No. 25GHYW009117
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl 3,3'-disulfanediyl(2R,2'R)-bis(2-aminopropanoate) dihydrochloride is a cysteine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 175 mg/mL (512.78 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.