Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl 3,3'-Dithiopropionimidate Dihydrochloride

Catalog No. D2388100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2388100MG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. D2388100MG Supplier Sigma Aldrich Supplier No. D2388100MG
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Synthesis of glutathione-sensitive cross-linked polyethylenimine gene vectorPreparation of cyclodextrin-containing polymers designed for gene deliveryCrosslinking of chick oviduct progesterone-receptor subunitsNote that the amidine linkage preserves original primary amine positive charge. The disulfide linkage is cleavable by mild reduction. Incorporates an eight ato mLinker.

Specifications

CAS 868-59-7
Melting Point 123°C to 125°C
Molecular Formula C5H11NO2S -+ HCl
Linear Formula HSCH2CH(NH2)COOC2H5 -+ HCl
MDL Number MFCD00012631
Quantity 100 g
Molecular Weight (g/mol) 185.67 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.