Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 3,3'-Thiodipropionate 98.0+%, TCI America™
SDP

Catalog No. T0901500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0901500ML 500 mL
T090125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0901500ML Supplier TCI America Supplier No. T0901500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4131-74-2
Molecular Formula C8H14O4S
Molecular Weight (g/mol) 206.256
MDL Number MFCD00008465
InChI Key MYWWWNVEZBAKHR-UHFFFAOYSA-N
Synonym 3,3′C-Thiodipropionic Acid Dimethyl Ester
PubChem CID 77781
IUPAC Name methyl 3-(3-methoxy-3-oxopropyl)sulfanylpropanoate
SMILES COC(=O)CCSCCC(=O)OC

Specifications

Color Yellow
Boiling Point 164°C
Quantity 500 mL
Formula Weight 206.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl 3,3′-Thiodipropionate
EINECSNumber (2)-1398
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.