Learn More
Sigma Aldrich DIMETHYL 3-METHYL-5-(1H-PYRROL-1-YL)THIOPHENE-2 4-DICARBOXYLATE
Catalog No. TMT005411G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
TMT005411G 5 g
1 options
Catalog No. TMT005411G Supplier Sigma Aldrich Supplier No. TMT005411G
  • NEW LOWER PRICE
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
The focal 1,8-naphthalimide fluorophores in the antennae were modified with 1,2,2,6,6-pentamethylpiperidine to improve their photostability.1,2,2,6,6-Pentamethylpiperidine (PMP) was used as an organic structure directing agent (OSDA) in the synthesis of the RTH-type zeolites.
Specifications
Quantity 5 g