Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 4,4'-Biphenyldicarboxylate 98.0+%, TCI America™
SDP

Catalog No. B13095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13095G 5 g
B130925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13095G Supplier TCI America Supplier No. B13095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 792-74-5
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.28
MDL Number MFCD00017201
InChI Key BKRIRZXWWALTPU-UHFFFAOYSA-N
Synonym dimethyl 1,1'-biphenyl-4,4'-dicarboxylate, dimethyl biphenyl-4,4'-dicarboxylate, biphenyl dimethyl dicarboxylate, dimethyl 4,4'-biphenyldicarboxylate, 1,1'-biphenyl-4,4'-dicarboxylic acid, dimethyl ester, unii-k61bxa0u9c, 4,4'-biphenyldicarboxylic acid, dimethyl ester, k61bxa0u9c, methyl 4-4-methoxycarbonylphenyl benzoate, methyl 4-4-methoxycarbonyl phenyl benzoate
PubChem CID 13099
IUPAC Name 4,4'-dimethyl [1,1'-biphenyl]-4,4'-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 217°C
Color White
Quantity 5 g
Formula Weight 270.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 4,4′-Biphenyldicarboxylate
EINECSNumber (4)-0832
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.