Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 4-Chloro-2,6-pyridinedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. D46931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46931G Supplier TCI America Supplier No. D46931G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent with >98.0% purity as determined by GC.

Chemical Identifiers

CAS 5371-70-0
Molecular Formula C9H8ClNO4
Molecular Weight (g/mol) 229.616
MDL Number MFCD02612781
InChI Key NFXKYKHKNUFOKB-UHFFFAOYSA-N
Synonym dimethyl 4-chloro-2,6-pyridinedicarboxylate, methyl 4-chloropyridine-2,6-dicarboxylate, 2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate, 2,6-pyridinedicarboxylic acid, 4-chloro-, dimethyl ester, 4-chloropyridine-2,6-dicarboxylic acid,dimethyl ester, methyl 4-chloro-6-methoxycarbonyl pyridine-2-carboxylate, 4-chloro-2,6-pyridinedicarboxylic acid dimethyl ester, pubchem20951, pych37, ksc496e3d
PubChem CID 237845
IUPAC Name dimethyl 4-chloropyridine-2,6-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=N1)C(=O)OC)Cl

Specifications

Melting Point 142°C
Quantity 1 g
Formula Weight 229.62
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 4-Chloro-2,6-pyridinedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.