Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 4-chloropyridine-2,6-dicarboxylate, 97%

Catalog No. AAH6484106
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6484106 5 g
AAH64841MD 250 mg
AAH6484103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6484106 Supplier Thermo Scientific Chemicals Supplier No. H6484106
Only null left
Add to Cart

CAS: 5371-70-0 | C9H8ClNO4 | 229.616 g/mol

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5371-70-0
Molecular Formula C9H8ClNO4
Molecular Weight (g/mol) 229.616
MDL Number MFCD02612781
InChI Key NFXKYKHKNUFOKB-UHFFFAOYSA-N
Synonym dimethyl 4-chloro-2,6-pyridinedicarboxylate, methyl 4-chloropyridine-2,6-dicarboxylate, 2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate, 2,6-pyridinedicarboxylic acid, 4-chloro-, dimethyl ester, 4-chloropyridine-2,6-dicarboxylic acid,dimethyl ester, methyl 4-chloro-6-methoxycarbonyl pyridine-2-carboxylate, 4-chloro-2,6-pyridinedicarboxylic acid dimethyl ester, pubchem20951, pych37, ksc496e3d
PubChem CID 237845
IUPAC Name dimethyl 4-chloropyridine-2,6-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=N1)C(=O)OC)Cl

Specifications

Melting Point 139°C to 144°C
Color Yellow
Quantity 5 g
Beilstein 197075
Solubility Information Slightly soluble in water.
Formula Weight 229.62
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Dimethyl 4-chloropyridine-2,6-dicarboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.