Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 5-nitroisophthalate, 98+%

Catalog No. AAA106100B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA106100B 1000 g
AAA1061030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA106100B Supplier Thermo Scientific Chemicals Supplier No. A106100B
Only null left
Add to Cart

CAS: 13290-96-5 | C10H9NO6 | 239.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13290-96-5
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.18
MDL Number MFCD00008429
InChI Key GGTSJKFPGKFLCZ-UHFFFAOYSA-N
Synonym dimethyl 5-nitroisophthalate, 5-nitroisophthalic acid dimethyl ester, 5-nitroisophthalic acid, dimethyl ester, 1,3-benzenedicarboxylic acid, 5-nitro-, dimethyl ester, dimethyl 5-nitro-1,3-benzenedicarboxylate, isophthalic acid, 5-nitro-, dimethyl ester, 1,3-dimethyl 5-nitrobenzene-1,3-dicarboxylate, 5-nitro dimethyl isophthalate, 5-nitro-1,3-benzenedicarboxylic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-nitro-, 1,3-dimethyl ester
PubChem CID 83316
IUPAC Name dimethyl 5-nitrobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)[N+]([O-])=O

Specifications

Melting Point 121°C to 123°C
Quantity 1000 g
Beilstein 2140916
Formula Weight 239.19
Percent Purity ≥98%
Chemical Name or Material Dimethyl 5-nitroisophthalate
EINECSNumber 236-307-3
RTECSNumber CZ4340000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.