Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 5-Nitroisophthalate, TCI America™
SDP

Catalog No. N056725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N056725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N056725G Supplier TCI America Supplier No. N056725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13290-96-5
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.18
MDL Number MFCD00008429
InChI Key GGTSJKFPGKFLCZ-UHFFFAOYSA-N
Synonym dimethyl 5-nitroisophthalate, 5-nitroisophthalic acid dimethyl ester, 5-nitroisophthalic acid, dimethyl ester, 1,3-benzenedicarboxylic acid, 5-nitro-, dimethyl ester, dimethyl 5-nitro-1,3-benzenedicarboxylate, isophthalic acid, 5-nitro-, dimethyl ester, 1,3-dimethyl 5-nitrobenzene-1,3-dicarboxylate, 5-nitro dimethyl isophthalate, 5-nitro-1,3-benzenedicarboxylic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-nitro-, 1,3-dimethyl ester
PubChem CID 83316
IUPAC Name 1,3-dimethyl 5-nitrobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)C(=O)OC)[N+]([O-])=O

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 25 g
Formula Weight 239.18
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 5-Nitroisophthalate
RTECSNumber CZ4340000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.