Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Dimethyl acetylenedicarboxylate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Versatile dienophile used in Diels-Alder reactions.
Specifications
Specifications
| CAS | 762-42-5 |
| Density | 1.156 g/mL (at 25°C (literature)) |
| Boiling Point | 95°C to 98°C (19 mmHg) |
| Molecular Formula | C6H6O4 |
| Refractive Index | n20/D 1.447 (literature) |
| Linear Formula | CH3OCOC///CCO2CH3 |
| MDL Number | MFCD00008456 |
| Quantity | 5 g |
| Synonym | Dimethyl 2-butynedioate |
| Molecular Weight (g/mol) | 142.11 |
| Afficher plus de résultats |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'