Learn More
Medchemexpress LLC Dimethyl adipate | 627-93-0 | MFCD00008469 | 100.0% | 174.19 g·mol⁻¹ | C8H14O4 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW015301500G
Encompass_Preferred
Dimethyl adipate (hexanedioic acid dimethyl ester) is an aliphatic dicarboxylate ester commonly used as a plasticizer and as an intermediate in organic synthesis. It is supplied in laboratory and industrial quantities with high stated purity and is suitable for formulation, polymer modification, and laboratory-scale synthesis.
  • High purity suitable for research and analytical use.
  • Acts as an effective plasticizer for polymers and formulations.
  • Useful intermediate in organic synthesis and esterification reactions.
  • Low volatility and good solvent compatibility for processing.
  • Available in multiple laboratory sizes for scale flexibility.
  • Clear liquid that is easy to handle and incorporate into formulations.
Catalog No. 50-003-02926